Merck
  • Comparison of different sorbents for multiresidue solid-phase extraction of 16 pesticides from groundwater coupled with high-performance liquid chromatography.
Comparison of different sorbents for multiresidue solid-phase extraction of 16 pesticides from groundwater coupled with high-performance liquid chromatography.
Talanta (2007-01-15)
Angelo Antonio D'Archivio, Maria Fanelli, Pietro Mazzeo, Fabrizio Ruggieri
ABSTRACT
A procedure based on solid-phase extraction (SPE) followed by high-performance liquid chromatography (HPLC) with diode array detection has been developed for the simultaneous analysis of 16 widely used pesticides in groundwater samples. The compounds analysed were: aldicarb, atrazine, desethylatrazine, desysopropylatrazine, carbofuran, 2,4-D, dicloran, fenitrothion, iprodione, linuron, metalaxyl, metazachlor, phenmedipham, procymidone, simazine and vinclozolin. Five different SPE sorbents, C(18) bonded silica (Isolute SPE C18 (EC)), graphitised carbon black (Superclean Envi-Carb), highly cross-linked polystyrene-divinylbenzene (Lichrolut EN), divinylbenzene-N-vinylpyrrolidone (Oasis HLB) and surface modified styrene-divinylbenzene (Strata X), were compared. HPLC separation and quantification of the selected pesticides was carried out under isocratic conditions by means of a new reversed-phase column (Gemini from Phenomenex) based on C(18) bonded to organic-silica particles. Oasis HLB and Strata X provided the best results in the preconcentration of 1-l samples, yielding average recoveries higher than 70%, except for phenmedipham that rapidly degrades in groundwater. Detection limits of the target pesticides provided by the proposed SPE-HPLC procedure were between 0.003 and 0.04microg l(-1).
Product Description
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pack of 100, AOAC® 2007.01
Supelco
Supel QuE, PSA/ENVI-Carb Tube 1, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb Tube 2, pk of 50, suitable for EN 15662:2008 per BS, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 1, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/ENVI-Carb (EN) Cleanup Tube 2, pack of 100, suitable for EN 15662:2008 per BS
Supelco
Supel QuE, PSA/C18/ENVI-Carb Tube, pk of 50, AOAC® 2007.01, centrifuge tube volume 15 mL , Shaker Compatible