Merck
DISTAG/TBCCd1 Is Required for Basal Cell Fate Determination in
The Plant cell (2017-12-07)
Olivier Godfroy, Toshiki Uji, Chikako Nagasato, Agnieszka P Lipinska, Delphine Scornet, Akira F Peters, Komlan Avia, Sebastien Colin, Laure Mignerot, Taizo Motomura, J Mark Cock, Susana M Coelho
ABSTRACT
Brown algae are one of the most developmentally complex groups within the eukaryotes. As in many land plants and animals, their main body axis is established early in development, when the initial cell gives rise to two daughter cells that have apical and basal identities, equivalent to shoot and root identities in land plants, respectively. We show here that mutations in the
Sigma-Aldrich
MISSION® esiRNA, targeting human MAX