Merck
  • Analogues of Disulfides from Allium stipitatum Demonstrate Potent Anti-tubercular Activities through Drug Efflux Pump and Biofilm Inhibition.
Analogues of Disulfides from Allium stipitatum Demonstrate Potent Anti-tubercular Activities through Drug Efflux Pump and Biofilm Inhibition.
Scientific reports (2018-01-20)
Cynthia A Danquah, Eleftheria Kakagianni, Proma Khondkar, Arundhati Maitra, Mukhlesur Rahman, Dimitrios Evangelopoulos, Timothy D McHugh, Paul Stapleton, John Malkinson, Sanjib Bhakta, Simon Gibbons
ABSTRACT
Disulfides from Allium stipitatum, commonly known as Persian shallot, were previously reported to possess antibacterial properties. Analogues of these compounds, produced by S-methylthiolation of appropriate thiols using S-methyl methanethiosulfonate, exhibited antimicrobial activity, with one compound inhibiting the growth of Mycobacterium tuberculosis at 17 µM (4 mg L
Sigma-Aldrich
7-Fluorobenzo[d]thiazole-2-thiol, AldrichCPR
Sigma-Aldrich
Aldrithiol-4, 98%