Merck
  • Synergistic Effect of Graphene Oxide Coated Nanotised Apigenin with Paclitaxel (GO-NA/PTX): A ROS Dependent Mitochondrial Mediated Apoptosis in Ovarian Cancer.
Synergistic Effect of Graphene Oxide Coated Nanotised Apigenin with Paclitaxel (GO-NA/PTX): A ROS Dependent Mitochondrial Mediated Apoptosis in Ovarian Cancer.
Anti-cancer agents in medicinal chemistry (2017-04-27)
Manish Kumar Pal, Shyam Pyari Jaiswar, Ashish Dwivedi, Shruti Goyal, Vinay Nand Dwivedi, Anumesh Kumar Pathak, Vinod Kumar, Pushp Lata Sankhwar, Ratan Singh Ray
ABSTRACT
Ovarian cancer is most lethal among all gynecologic malignancies. Paclitaxel (PTX) is well used chemotherapeutic regimen for cancer control; however its undesired toxicity has been a matter of concern for clinicians. Here, we used the graphene oxide coated nanotised apigenin (GO-NA) to enhance the efficacy of paclitaxel. The combined use of paclitaxel (PTX) and nanotised apigenin (NA) may reduce the PTX dose and increase the efficacy. GO and GO-Apigenin was prepared by modified Hummers method and the nanoparticles were characterized by dynamic light scattering and transmission electron microscopy. SKOV-3 cells were treated by DMSO, Group I (Control)-McCoy's 5A Medium, Group II-Paclitaxel (5nM), Group III- Nanotised Apigenin (GO-NA-10µM), Group IV- Paclitaxel (5nM) + GO-NA (10µM). Cell viability and IC-50 value were determined by MTT assay, synergism by Compusyn software, ROS by DCFH-DA assay, SOD activity by kit and MMP were examined by JC-1 and mitotracker/DAPI staining, cell cycle by flow cytometry, mRNA and protein level by Real Time-PCR and Western blot respectively Results: Results showed that GO-NA-PTX enhanced the anti-proliferative effect in synergistic manner as compare to GO-NA and PTX alone. GO-NA-PTX significantly suppressed the SOD activity, promotes the ROS accumulation, mitochondrial depolarization, DNA integrity and cell cycle arrest collectively accord the apoptosis. Results of immunocytochemistry, RT-PCR and western blot showed up-regulation of caspase-3, Bax, and down-regulation of Bcl-2. The combination of PTX with GO-NA produces synergistic effects in SKOV-3 cells via the modulation of pro and anti-apoptotic gene and may reduce side effects of PTX.
Sigma-Aldrich
Sodium oxide, 80%
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
N-(3-Methoxypropyl)acrylamide, contains MEHQ as inhibitor, 95%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)