- New chemical synthesis of functionalized arylzinc compounds from aromatic or thienyl bromides under mild conditions using a simple cobalt catalyst and zinc dust.
New chemical synthesis of functionalized arylzinc compounds from aromatic or thienyl bromides under mild conditions using a simple cobalt catalyst and zinc dust.
Journal of the American Chemical Society (2003-03-27)
Hyacinthe Fillon, Corinne Gosmini, Jacques Périchon
PMID12656620
ABSTRACT
A new chemical method for the preparation of arylzinc intermediates is described in acetonitrile, on the basis of the activation of aryl bromides by low-valent cobalt species arising from the reduction of cobalt halide by zinc dust. This procedure allows for the synthesis of a variety of functionalized aryl- and thienylzinc species in good to excellent yields. The versatility and the simplicity of that original method represent an alternative to most known procedures.
MATERIALS