Merck
  • Acquired vorinostat resistance shows partial cross-resistance to 'second-generation' HDAC inhibitors and correlates with loss of histone acetylation and apoptosis but not with altered HDAC and HAT activities.
Acquired vorinostat resistance shows partial cross-resistance to 'second-generation' HDAC inhibitors and correlates with loss of histone acetylation and apoptosis but not with altered HDAC and HAT activities.
Anti-cancer drugs (2009-03-27)
Konstantin J Dedes, Ioannis Dedes, Patrick Imesch, André O von Bueren, Daniel Fink, André Fedier
ABSTRACT
Histone deacetylase (HDAC) inhibitors such as vorinostat (suberoylanilide hydroxamic acid), valproic acid, romidepsin (FK-228), and LBH589 comprise a relatively new class of potent anticancer agents. This study provides evidence for the potential of vorinostat to cause acquisition of multidrug resistance protein-independent resistance in HCT116 colon tumor cells. This acquired resistance is moderate (two-fold to three-fold), is nonreversible, and correlates with the loss of responses typically seen with HDAC inhibitors, that is the loss of acetylation of the histones H2A, H2B, H3, and H4, the loss of the G2/M checkpoint activation, and the loss of caspase 3-dependent and caspase 7-dependent apoptosis. This acquired resistance also associates with cross-resistance to the hydroxamate-class (LBH589 and JNJ26481585) and to the aliphatic acid-class (valproic acid) HDAC inhibitors but not to the benzamide-class (MGCD0103) and the cyclic peptide-class (romidepsin) HDAC inhibitors. The acquired HDAC inhibitor resistance described hereis not a result of altered HDAC and histone acetyltransferase activities and differs from that previously reported for romidepsin.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-HDAC2 Antibody, clone 3F3, clone 3F3, Upstate®, from mouse
Sigma-Aldrich
Anti-HDAC3 Antibody, clone 3G6, clone 3G6, Upstate®, from mouse
Sigma-Aldrich
Anti-HDAC6 Antibody, CT, Upstate®, from rabbit