Merck
ChaC2, an Enzyme for Slow Turnover of Cytosolic Glutathione.
The Journal of biological chemistry (2016-12-04)
Amandeep Kaur, Ruchi Gautam, Ritika Srivastava, Avinash Chandel, Akhilesh Kumar, Subramanian Karthikeyan, Anand Kumar Bachhawat
ABSTRACT
Glutathione degradation plays an important role in glutathione and redox homeostasis, and thus it is imperative to understand the enzymes and the mechanisms involved in glutathione degradation in detail. We describe here ChaC2, a member of the ChaC family of γ-glutamylcyclotransferases, as an enzyme that degrades glutathione in the cytosol of mammalian cells. ChaC2 is distinct from the previously described ChaC1, to which ChaC2 shows ∼50% sequence identity. Human and mouse ChaC2 proteins purified in vitro show 10-20-fold lower catalytic efficiency than ChaC1, although they showed comparable K
Sigma-Aldrich
Anti-CHAC2, (N-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CHAC1 antibody produced in rabbit, affinity isolated antibody