Merck
  • Identity and Diversity of Human Peripheral Th and T Regulatory Cells Defined by Single-Cell Mass Cytometry.
Identity and Diversity of Human Peripheral Th and T Regulatory Cells Defined by Single-Cell Mass Cytometry.
Journal of immunology (Baltimore, Md. : 1950) (2017-11-29)
Matthew A Kunicki, Laura C Amaya Hernandez, Kara L Davis, Rosa Bacchetta, Maria-Grazia Roncarolo
ABSTRACT
Human CD3
Sigma-Aldrich
AK-7, ≥98% (HPLC)