Merck
  • Evaluation of inositol phosphates in urine after topical administration of myo-inositol hexaphosphate to female Wistar rats.
Evaluation of inositol phosphates in urine after topical administration of myo-inositol hexaphosphate to female Wistar rats.
Life sciences (2017-11-21)
F Grases, A Costa-Bauzá, F Berga, A Rodríguez, R M Gomila, G Martorell, M R Martínez-Cignoni
ABSTRACT
Previous studies demonstrated a remarkable increase of urinary InsP We fed 12 female rats with a diet without InsP At baseline, after dietary deprivation of InsP We conclude that the skin can absorb InsP
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.), 8.74% (ZN (THEORY))