Merck
  • Crystal and NMR Structures of a Peptidomimetic β-Turn That Provides Facile Synthesis of 13-Membered Cyclic Tetrapeptides.
Crystal and NMR Structures of a Peptidomimetic β-Turn That Provides Facile Synthesis of 13-Membered Cyclic Tetrapeptides.
Chemistry, an Asian journal (2017-11-04)
Alan J Cameron, Christopher J Squire, Patrick J B Edwards, Elena Harjes, Vijayalekshmi Sarojini
ABSTRACT
Herein we report the unique conformations adopted by linear and cyclic tetrapeptides (CTPs) containing 2-aminobenzoic acid (2-Abz) in solution and as single crystals. The crystal structure of the linear tetrapeptide H
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
2,3,5-Collidine, 99%
Sigma-Aldrich
1,3-Propylene sulfite, 99%