Merck
  • CD81 association with SAMHD1 enhances HIV-1 reverse transcription by increasing dNTP levels.
CD81 association with SAMHD1 enhances HIV-1 reverse transcription by increasing dNTP levels.
Nature microbiology (2017-09-06)
Vera Rocha-Perugini, Henar Suárez, Susana Álvarez, Soraya López-Martín, Gina M Lenzi, Felipe Vences-Catalán, Shoshana Levy, Baek Kim, María A Muñoz-Fernández, Francisco Sánchez-Madrid, Maria Yáñez-Mó
ABSTRACT
In this study, we report that the tetraspanin CD81 enhances human immunodeficiency virus (HIV)-1 reverse transcription in HIV-1-infected cells. This is enabled by the direct interaction of CD81 with the deoxynucleoside triphosphate phosphohydrolase SAMHD1. This interaction prevents endosomal accumulation and favours the proteasome-dependent degradation of SAMHD1. Consequently, CD81 depletion results in SAMHD1 increased expression, decreasing the availability of deoxynucleoside triphosphates (dNTP) and thus HIV-1 reverse transcription. Conversely, CD81 overexpression, but not the expression of a CD81 carboxy (C)-terminal deletion mutant, increases cellular dNTP content and HIV-1 reverse transcription. Our results demonstrate that the interaction of CD81 with SAMHD1 controls the metabolic rate of HIV-1 replication by tuning the availability of building blocks for reverse transcription, namely dNTPs. Together with its role in HIV-1 entry and budding into host cells, the data herein indicate that HIV-1 uses CD81 as a rheostat that controls different stages of the infection.
Sigma-Aldrich
MISSION® esiRNA, targeting human SAMHD1
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-glycerol, ≥95% (TLC), liquid
Sigma-Aldrich
MISSION® esiRNA, targeting human CD81