Merck
  • Platelet rescue by macrophage depletion in obese ADAMTS-13-deficient mice at risk of thrombotic thrombocytopenic purpura.
Platelet rescue by macrophage depletion in obese ADAMTS-13-deficient mice at risk of thrombotic thrombocytopenic purpura.
Journal of thrombosis and haemostasis : JTH (2017-11-10)
L Geys, E Roose, I Scroyen, H Rottensteiner, C Tersteeg, M F Hoylaerts, K Vanhoorelbeke, H R Lijnen
ABSTRACT
Essentials Obesity is a potential risk factor for development of thrombotic thrombocytopenic purpura (TTP). Obese ADAMTS-13-deficient mice were triggered with von Willebrand factor (VWF). Depletion of hepatic and splenic macrophages protects against thrombocytopenia in this model. VWF enhances phagocytosis of platelets by macrophages, dose-dependently. Background Thrombotic thrombocytopenic purpura (TTP) is caused by the absence of ADAMTS-13 activity. Thrombocytopenia is presumably related to the formation of microthrombi rich in von Willebrand factor (VWF) and platelets. Obesity may be a risk factor for TTP; it is associated with abundance of macrophages that may phagocytose platelets. Objectives To evaluate the role of obesity and ADAMTS-13 deficiency in TTP, and to establish whether macrophages contribute to thrombocytopenia. Methods Lean or obese ADAMTS-13-deficient (Adamts-13
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder