Merck
  • Catalase ameliorates diabetes-induced cardiac injury through reduced p65/RelA- mediated transcription of BECN1.
Catalase ameliorates diabetes-induced cardiac injury through reduced p65/RelA- mediated transcription of BECN1.
Journal of cellular and molecular medicine (2017-06-24)
Xu Wang, Youli Tao, Yewei Huang, Kungao Zhan, Mei Xue, Ying Wang, Dandan Ruan, Yangzhi Liang, Xiaozhong Huang, Jianjun Lin, Zhiwei Chen, Lingchun Lv, Santie Li, Gen Chen, Yang Wang, Ruijie Chen, Weitao Cong, Litai Jin
ABSTRACT
Catalase is an antioxidative enzyme that converts hydrogen peroxide (H
Sigma-Aldrich
MISSION® esiRNA, targeting human RELA