Merck
  • Direct selection of Pichia pastoris expression strains using new G418 resistance vectors.
Direct selection of Pichia pastoris expression strains using new G418 resistance vectors.
Yeast (Chichester, England) (2008-03-11)
Joan Lin-Cereghino, Matthew D Hashimoto, Allison Moy, James Castelo, Claire C Orazem, Peter Kuo, See Xiong, Vishal Gandhi, Christopher T Hatae, Alex Chan, Geoff P Lin-Cereghino
ABSTRACT
The methylotrophic yeast, Pichia pastoris, is widely used as a host organism for the expression of heterologous proteins. Currently, the Zeocin and blasticidin resistance genes are the only dominant selectable markers that can be used for primary selection of transformants. In this report we describe new expression vectors that can be used to select directly for P. pastoris transformants using G418 resistance conferred by a modified Tn903kan(r) gene. Compared to other dominant markers, this system is more economical and offers a higher transformation efficiency, due to the small sizes of the cloning vectors, pKAN B and pKANalpha B (GenBank Accession Nos EU285585 and EU285586, respectively). Additionally, multicopy transformants can be generated using these new vectors.
Sigma-Aldrich
G 418 disulfate salt solution, 50 mg/mL in H2O, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture