Merck
  • Potassium Regulation in Medaka (Oryzias latipes) Larvae Acclimated to Fresh Water: Passive Uptake and Active Secretion by the Skin Cells.
Potassium Regulation in Medaka (Oryzias latipes) Larvae Acclimated to Fresh Water: Passive Uptake and Active Secretion by the Skin Cells.
Scientific reports (2017-11-28)
Jiun-Lin Horng, Li-Lu Yu, Sian-Tai Liu, Po-Yen Chen, Li-Yih Lin
ABSTRACT
Molecular mechanisms of Na
Sigma-Aldrich
Chlorodimethylsilane, 98%
Sigma-Aldrich
2,4,6-Triaminopyrimidine, 97%
Sigma-Aldrich
VU591, ≥98% (HPLC)
Sigma-Aldrich
Ethyl 3-aminobenzoate, 97%
Ouabain, European Pharmacopoeia (EP) Reference Standard