- Magnesium homeostasis protects Salmonella against nitrooxidative stress.
Magnesium homeostasis protects Salmonella against nitrooxidative stress.
Scientific reports (2017-11-10)
Travis J Bourret, Lin Liu, Jeff A Shaw, Maroof Husain, Andrés Vázquez-Torres
PMID29118452
ABSTRACT
The PhoPQ two-component regulatory system coordinates the response of Salmonella enterica serovar Typhimurium to diverse environmental challenges encountered during infection of hosts, including changes in Mg