Merck
  • Bis(Cyclic Alkyl Amino Carbene) Ruthenium Complexes: A Versatile, Highly Efficient Tool for Olefin Metathesis.
Bis(Cyclic Alkyl Amino Carbene) Ruthenium Complexes: A Versatile, Highly Efficient Tool for Olefin Metathesis.
Angewandte Chemie (International ed. in English) (2016-12-13)
Rafał Gawin, Anna Kozakiewicz, Piotr A Guńka, Paweł Dąbrowski, Krzysztof Skowerski
ABSTRACT
The state-of-the-art in olefin metathesis is application of N-heterocyclic carbene (NHC)-containing ruthenium alkylidenes for the formation of internal C=C bonds and of cyclic alkyl amino carbene (CAAC)-containing ruthenium benzylidenes in the production of terminal olefins. A straightforward synthesis of bis(CAAC)Ru indenylidene complexes, which are highly effective in the formation of both terminal and internal C=C bonds at loadings as low as 1 ppm, is now reported.
Sigma-Aldrich
UltraCat, ≥95%