Merck
  • Lactococcus lactis LM0230 contains a single aminotransferase involved in aspartate biosynthesis, which is essential for growth in milk.
Lactococcus lactis LM0230 contains a single aminotransferase involved in aspartate biosynthesis, which is essential for growth in milk.
Microbiology (Reading, England) (2001-02-13)
E Dudley, J Steele
ABSTRACT
Amino acid aminotransferases (ATases), which catalyse the last biosynthetic step of many amino acids, may have important physiological functions in Lactococcus lactis during growth in milk. In this study, the aspartate ATase gene (aspC) from L. lactis LM0230 was cloned by complementation into Escherichia coli DL39. One chromosomal fragment putatively encoding aspC was partially sequenced. A 1179 bp ORF was identified which could encode for a 393 aa, 43.2 kDa protein. The deduced amino acid sequence had high identity to other AspC sequences in GenBank and is a member of the Igamma family of ATases. Substrate-specificity studies suggested that the lactococcal AspC has ATase activity only with aspartic acid (Asp). An internal deletion was introduced into the L. lactis chromosomal copy of aspC by homologous recombination. The wild-type and mutant strain grew similarly in defined media containing all 20 amino acids and did not grow in minimal media unless supplemented with asparagine (Asn). The mutant strain was also unable to grow in or significantly acidify milk unless supplemented with Asp or Asn. These results suggest that only one lactococcal ATase is involved in the conversion of oxaloacetate to Asp, and Asp biosynthesis is required for the growth of L. lactis LM0230 in milk.
Product Description
G-CAPSULE electroelution device
G-CAPSULE Accessory Weight