Merck
  • Chryseobacterium ginsengiterrae sp. nov., with Beta-Glucosidase Activity Isolated from Soil of a Ginseng Field.
Chryseobacterium ginsengiterrae sp. nov., with Beta-Glucosidase Activity Isolated from Soil of a Ginseng Field.
Current microbiology (2017-08-22)
Jong-Hun Noh, Van-An Hoang, Yeon-Ju Kim, Jong-Pyo Kang, Deok-Chun Yang
ABSTRACT
The isolated Chryseobacterium ginsengiterrae sp. nov DCY68
Sigma-Aldrich
1,8-Diaminooctane, 98%