Merck
  • Src-dependent phosphorylation of μ-opioid receptor at Tyr336 modulates opiate withdrawal.
Src-dependent phosphorylation of μ-opioid receptor at Tyr336 modulates opiate withdrawal.
EMBO molecular medicine (2017-08-19)
Lei Zhang, Cherkaouia Kibaly, Yu-Jun Wang, Chi Xu, Kyu Young Song, Patrick W McGarrah, Horace H Loh, Jing-Gen Liu, Ping-Yee Law
ABSTRACT
Opiate withdrawal/negative reinforcement has been implicated as one of the mechanisms for the progression from impulsive to compulsive drug use. Increase in the intracellular cAMP level and protein kinase A (PKA) activities within the neurocircuitry of addiction has been a leading hypothesis for opiate addiction. This increase requires the phosphorylation of μ-opioid receptor (MOR) at Tyr336 by Src after prolonged opiate treatment in vitro Here, we report that the Src-mediated MOR phosphorylation at Tyr336 is a prerequisite for opiate withdrawal in mice. We observed the recruitment of Src in the vicinity of MOR and an increase in phosphorylated Tyr336 (pY336) levels during naloxone-precipitated withdrawal. The intracerebroventricular or stereotaxic injection of a Src inhibitor (AZD0530), or Src shRNA viruses attenuated pY336 levels, and several somatic withdrawal signs. This was also observed in Fyn-/- mice. The stereotaxic injection of wild-type MOR, but not mutant (Y336F) MOR, lentiviruses into the locus coeruleus of MOR-/- mice restored somatic withdrawal jumping. Regulating pY336 levels during withdrawal might be a future target for drug development to prevent opiate addictive behaviors.
Sigma-Aldrich
Anti-phospho-Src (Tyr416) Antibody, clone 9A6, clone 9A6, Upstate®, from mouse
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)