Merck
  • PDGF/VEGF-Related Receptor Affects Transglutaminase Activity to Control Cell Migration During Crustacean Hematopoiesis.
PDGF/VEGF-Related Receptor Affects Transglutaminase Activity to Control Cell Migration During Crustacean Hematopoiesis.
Stem cells and development (2017-08-15)
Kingkamon Junkunlo, Kenneth Söderhäll, Chadanat Noonin, Irene Söderhäll
ABSTRACT
The platelet-derived growth factor (PDGF) receptor, a tyrosine kinase (TK) receptor whose ligand is PDGF, is crucial in the transduction of extracellular signals into cells and mediates numerous processes, such as cell proliferation, differentiation, survival, and migration. We demonstrate the important roles of a receptor TK related to the PDGF/VEGF family protein (PVR) in controlling hematopoietic progenitor cell migration by affecting extracellular transglutaminase (TGase) activity. Pl_PVR1, GenBank accession No. KY444650, is highly expressed in hemocytes and the hematopoietic tissue (HPT). Sunitinib malate was used to block the PVF/PVR downstream pathway in HPT cell culture. The addition of Sunitinib also caused the HPT cells to increase in size and begin spreading. An increase in extracellular TGase activity on the HPT cell membrane was observed in a dose-dependent manner after treatment with Sunitinib malate. The presence of crude Ast1 provided a combinatorial beneficial effect that enhanced the number of spreading cells after inhibition of the Pl_PVR downstream signaling cascade. In addition, an increased immunoreactivity for β-tubulin and elongation of β-tubulin filaments were found in Pl_PVR signaling-inhibited cells. The potential roles of PVF/PVR signaling in controlling progenitor cell activity during hematopoiesis in crayfish were investigated and discussed.
Sigma-Aldrich
Sunitinib malate, ≥98% (HPLC)