Merck
Persulfate enhanced photocatalytic degradation of bisphenol A by g-C
Chemosphere (2017-09-22)
Bochuan Liu, Meng Qiao, Yanbin Wang, Lijuan Wang, Yan Gong, Tao Guo, Xu Zhao
ABSTRACT
The enhancement of g-C
Sigma-Aldrich
2,4-Diamino-6-phenyl-1,3,5-triazine, 97%
Pricing and availability is not currently available.