Merck
  • Quantification and speciation of volatile fatty acids in the aqueous phase.
Quantification and speciation of volatile fatty acids in the aqueous phase.
Environmental pollution (Barking, Essex : 1987) (2017-06-27)
Jechan Lee, Jieun Kim, Jeong-Ik Oh, Sang-Ryong Lee, Eilhann E Kwon
ABSTRACT
This study lays great emphasis on establishing a reliable analytical platform to quantify and specify volatile fatty acids (VFAs) in the aqueous phase by derivatizing VFAs into their corresponding alkyl esters via thermally-induced rapid esterification (only 10 s reaction time). To this end, reaction conditions for the thermally-induced rapid esterification are optimized. A volumetric ratio of 0.5 at 400 °C for VFA/methanol is identified as the optimal reaction conditions to give ∼90% volatile fatty acid methyl ester (VFAME) yield. To maintain a high yield of VFAMEs, this study suggests that dilution of the sample to an optimum concentration (∼500 ppm for each VFA) is required. Derivatization of VFAs into VFAMEs via the thermally-induced rapid esterification is more reliable to quantify and specify VFAs in the aqueous phase than conventional colorimetric method.
Sigma-Aldrich
Isovaleric acid, natural, ≥98%, FG
Pricing and availability is not currently available.