Merck
A Novel Photocatalytic Purification System for Fish Culture.
Zebrafish (2017-07-06)
Basilio Randazzo, Giulia Chemello, Ivan Tortarolo, Gian Luca Chiarello, Maciej Zalas, Alessandra Santini, Margaret Liberatore, Marco Liberatore, Elena Selli, Ike Olivotto
ABSTRACT
The accumulation of nitrogen compounds represents a pivotal problem in the management of fish culture. Several methods were investigated in the last decades for treatment of waste waters, and the use of photocatalytic materials has received increasing attention. The photocatalytic degradation (PCD) process with titanium dioxide (TiO
Sigma-Aldrich
Indophenol Blue, Dye content 60 %