- Identification of claudin-4 binder that attenuates tight junction barrier function by TR-FRET-based screening assay.
Identification of claudin-4 binder that attenuates tight junction barrier function by TR-FRET-based screening assay.
Claudins are key functional and structural components of tight junctions (TJs) in epithelial cell sheets. The C-terminal fragment of Clostridium perfringens enterotoxin (C-CPE) binds to claudin-4 and reversibly modulates intestinal TJ seals, thereby enhancing paracellular transport of solutes. However, the use of C-CPE as an absorption enhancer is limited by the molecule's immunogenicity and manufacturing cost. Here, we developed a high-throughput screening system based on the Time-Resolved Fluorescence Resonance Energy Transfer (TR-FRET) method to identify claudin-4 binders in a library collection of 32,560 compounds. Thiostrepton, identified from the screen, decreased transepithelial electrical resistance and increased flux of 4-kDa fluorescein isothiocyanate-labelled dextran (FD-4) in Caco-2 cell monolayers, a model of intestinal epithelium. Thiostrepton changed the expression, but not the localisation, of TJ components. Treatment of rat jejunum with thiostrepton increased the absorption of FD-4 without tissue toxicity, indicating that thiostrepton is a novel claudin-4 binder that enhances intestinal permeability. The screening system may therefore be a useful tool for identifying claudin-4 binders to enhance drug absorption in mucosa.