Merck
  • Natural polymeric 3-alkylpyridinium salt affects vertebrate skeletal muscle contractility by preferentially blocking neuromuscular transmission.
Natural polymeric 3-alkylpyridinium salt affects vertebrate skeletal muscle contractility by preferentially blocking neuromuscular transmission.
Toxicology letters (2017-09-25)
Monika Cecilija Žužek, Marjana Grandič, Evelyne Benoit, Robert Frangež
ABSTRACT
The effects of natural polymeric alkylpyridinium salt (nPoly-3-APS), a potent acetylcholinesterase inhibitor isolated from the marine sponge Reniera sarai, were studied on isolated mouse phrenic nerve-hemidiaphragm muscle preparations using electrophysiological approaches. nPoly-3-APS inhibited nerve-evoked isometric muscle twitch and tetanic contraction in a concentration-dependent manner (IC
Sigma-Aldrich
3,4-Diaminopyridine, ≥98%