- Microheterogeneity in CH
Microheterogeneity in CH
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-07-29)
Władysław Wrzeszcz, Sylwester Mazurek, Roman Szostak, Paweł Tomza, Mirosław A Czarnecki
PMID28753528
ABSTRACT
Recently, we demonstrated the presence of microheterogeneity in binary mixtures of unlike alcohols. [RSC Adv. 2016, 6, 37195-37202] The aim of this work was examination if this phenomenon occurs also in the mixture of very similar alcohols like CH