Merck
  • CBP501 inhibits EGF-dependent cell migration, invasion and epithelial-to-mesenchymal transition of non-small cell lung cancer cells by blocking KRas to calmodulin binding.
CBP501 inhibits EGF-dependent cell migration, invasion and epithelial-to-mesenchymal transition of non-small cell lung cancer cells by blocking KRas to calmodulin binding.
Oncotarget (2017-11-02)
Naoya Saito, Naoki Mine, Donald W Kufe, Daniel D Von Hoff, Takumi Kawabe
ABSTRACT
The anti-cancer agent CBP501 binds to calmodulin (CaM). Recent studies showed that migration and metastasis are inhibited by several CaM antagonists. However, there is no available evidence that CBP501 has similar effects. Here we found that CBP501 inhibits migration of non-small cell lung cancer (NSCLC) cells
Sigma-Aldrich
PQ401, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human KRAS
Sigma-Aldrich
Calmidazolium chloride, solid