Merck
Nanoporous PtCo/Co
Journal of colloid and interface science (2017-09-05)
Qiuxia Zhou, Caixia Xu
ABSTRACT
The design and fabrication of highly active and durable catalysts for the ammonia borane (AB) hydrolysis has been one of significant issues in the application of green and economic hydrogen energy. Nanoporous (NP) PtCo/Co
Sigma-Aldrich
Borane-ammonia complex, 95%