Merck
  • Determination of 89 drugs and other micropollutants in unfiltered wastewater and freshwater by LC-MS/MS: an alternative sample preparation approach.
Determination of 89 drugs and other micropollutants in unfiltered wastewater and freshwater by LC-MS/MS: an alternative sample preparation approach.
Analytical and bioanalytical chemistry (2017-08-18)
Alexandros G Asimakopoulos, Pranav Kannan, Sean Higgins, Kurunthachalam Kannan
ABSTRACT
A liquid chromatography-triple quadrupole-tandem mass spectrometry (LC-qQq-MS/MS) method was developed for simultaneous determination of 89 legal neuropsychiatric pharmaceuticals and illicit drugs (both parent compounds and metabolites) and other micropollutants in unfiltered wastewater and freshwater. The target chemicals fall under the classes of amphetamine-type stimulants, cocaine compounds, opiates and opioids, benzodiazepines, lysergic compounds, antipsychotics, anesthetics, antiepileptics, antidepressants, sympathomimetics, cannabinoids, blood thinners, antihistamines, β-blockers, caffeine derivatives, nicotine derivatives, z-drugs, new designer drugs, and Alzheimer medications. The sample preparation procedure was designed for unfiltered wastewater and freshwater without the need to separate the particulate matter (if any) from the aqueous phase prior to extraction. Samples were pre-concentrated by rotary evaporation in the presence of a solvent. Method precision (absolute values; N = 6 replicate analyses at the fortification level of 50 ng, k = 6 days) for 87 out of 89 target analytes ranged from 2.8 to 34% (RSD %). The limits of detection ranged from 0.11 to 202 ng L
Product Description
Supelco
Atenolol-d7, analytical standard
Sigma-Aldrich
1,7-Dimethylxanthine, ~98%, solid