Merck
Preclinical Evaluation and Quantification of
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2016-10-30)
Shivashankar Khanapur, Aren van Waarde, Rudi A J O Dierckx, Philip H Elsinga, Michel J B Koole
ABSTRACT
The cerebral adenosine A
Sigma-Aldrich
1,3-Propanediol di-p-tosylate, 98%