Merck
  • Development and comparison of HPLC and MEKC methods for the analysis of cyclic sulfur mustard degradation products.
Development and comparison of HPLC and MEKC methods for the analysis of cyclic sulfur mustard degradation products.
Electrophoresis (2017-01-04)
Heidi Lees, Merike Vaher, Mihkel Kaljurand
ABSTRACT
In this study, novel, fast, and simple methods based on RP-HPLC and MEKC with DAD are developed and validated for the qualitative and quantitative determination of five cyclic sulfur mustard (HD) degradation products (1,4-thioxane, 1,3-dithiolane, 1,4-dithiane, 1,2,5-trithiepane, and 1,4,5-oxadithiepane) in water samples. The HPLC method employs a C18 column and an isocratic water-ACN (55:45, v/v) mobile phase. This method enables separation of all five cyclic compounds within 8 min. With the CE method, the baseline separation of five compounds was achieved in less than 11 min by applying a simple BGE composed of a 10 mM borate buffer and 90 mM SDS (pH 9.15). Both methods showed good linear correlation (R
Sigma-Aldrich
1,3-Dithiolane, 97%
Sigma-Aldrich
1,4-Thioxane, 98%