Merck
  • Bone Morphogenetic Protein-Modulator BMPER Regulates Endothelial Barrier Function.
Bone Morphogenetic Protein-Modulator BMPER Regulates Endothelial Barrier Function.
Inflammation (2016-12-21)
Thomas Helbing, Gwendoline Wiltgen, Alexandra Hornstein, Elena Z Brauers, Linus Arnold, Adrian Bauer, Jennifer S Esser, Philipp Diehl, Sebastian Grundmann, Katrin Fink, Cam Patterson, Christoph Bode, Martin Moser
ABSTRACT
The endothelium serves as a selective barrier and controls the exchange of nutrients, hormones, and leukocytes between blood and tissues. Molecular mechanisms contributing to the pathogenesis of endothelial barrier dysfunction remain incompletely understood. Accumulating evidence implicates bone morphogenetic protein (BMP)-modulator BMPER as a key regulator in endothelial biology. Herein, we analyze the impact of BMPER in the control of endothelial barrier function. To assess the role of BMPER in vascular barrier function in mice, we measured the leakage of Evans blue dye from blood into interstitial lung tissue. BMPER
Sigma-Aldrich
MISSION® esiRNA, targeting human BMPER
Sigma-Aldrich
MISSION® esiRNA, targeting human BMP4