- Nanodiamonds as Nucleating Agents for Protein Crystallization.
Nanodiamonds as Nucleating Agents for Protein Crystallization.
Langmuir : the ACS journal of surfaces and colloids (2017-06-13)
Yen-Wei Chen, Chien-Hsun Lee, Yung-Lin Wang, Tsung-Lin Li, Huan-Cheng Chang
PMID28602087
ABSTRACT
Nanodiamond (ND) is a carbon-based nanomaterial with potential for a wide range of biological applications. One of such applications is to facilitate the nucleation of protein crystals in aqueous solution. Here, we show that NDs (nominal diameters of 30 and 100 nm) after surface oxidation in air and subsequent treatment in strong acids are useful as heterogeneous nucleating agents for protein crystallization. Tested with lysozyme, ribonuclease A, proteinase K, and catalase, the nanomaterials in either aggregate or film form are found to be able to increase the crystallization efficiency of all proteins. Particularly, for 30 nm NDs, the films with an area of ∼2 mm
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein