Merck
Vibration and vibration-torsion levels of the S
The Journal of chemical physics (2017-07-03)
William D Tuttle, Adrian M Gardner, Laura E Whalley, Timothy G Wright
ABSTRACT
A study of the vibration and vibration-torsion levels of para-fluorotoluene in the 580-830 cm
Sigma-Aldrich
4-Fluorotoluene, 97%