Merck
  • Microencapsulation of Corn Wastewater (Nejayote) Phytochemicals by Spray Drying and Their Release Under Simulated Gastrointestinal Digestion.
Microencapsulation of Corn Wastewater (Nejayote) Phytochemicals by Spray Drying and Their Release Under Simulated Gastrointestinal Digestion.
Journal of food science (2017-06-10)
Javier Villela-Castrejón, Beatriz A Acosta-Estrada, Janet A Gutiérrez-Uribe
ABSTRACT
Corn lime cooking generates a large amount of wastewater known as nejayote that is composed of suspended solids and solubilized phytochemicals. Spray drying can be an alternative to recover bioactive molecules, such as ferulic acid, from nejayote. Besides the yield, the physicochemical properties (solubility, water activity, pH, moisture, hygroscopicity, total phenolic content, and distribution of free and bound hydroxycinnamic acids) of spray-dried nejayote powders were analyzed. The powders were obtained at 200 °C/100 °C or 150 °C/75 °C (inlet/outlet) air temperatures with the addition of maltodextrin (MD) or 2-hydroxypropyl-beta-cyclodextrin (HBCD) as encapsulating agents. Even when no carrier agent was used, a spray-dried nejayote powder was produced. The use of MD or HBCD as carrier increased the yield from 60.26% to 68.09% or 71.83%, respectively. As expected, a high inlet temperature (200 °C) allowed a satisfactory yield (>70%) and a low powder moisture (2.5%) desired by the industry. Water activity was reduced from 0.586 to 0.307 when HBCD was used in combination with a drying inlet temperature of 150 °C; and from 0.488 to 0.280 when the inlet temperature was set at 200 °C. Around 100% bioaccessibility of the compounds was observed after in vitro digestion. The addition of HBCD increased the release time (P < 0.05). Under simulated physiological conditions, there was no reduction of total phenolics, suggesting a good stability. This paper showed the feasibility to engage the spray drying technology to the corn industry to minimize their residues and reuse their by-products.
Sigma-Aldrich
Calconcarboxylic acid, suitable for use as an indicator for the titration of calcium