- Dansylglycine, a fluorescent probe for specific determination of halogenating activity of myeloperoxidase and eosinophil peroxidase.
Dansylglycine, a fluorescent probe for specific determination of halogenating activity of myeloperoxidase and eosinophil peroxidase.
Analytical biochemistry (2017-06-08)
Luiza de Carvalho Bertozo, Maria Luiza Zeraik, Valdecir Farias Ximenes
PMID28587811
ABSTRACT
Myeloperoxidase (MPO) and eosinophil peroxidase (EPO) are enzymes present in neutrophil and eosinophil leukocytes, respectively. Here, we present the development of a sensitive and specific assay for determination of the halogenating enzymatic activity of MPO and EPO based on the electrophilic attack of HOCl and HOBr on aromatic ring of dansylglycine (DG). We found that the intrinsic fluorescence of DG was promptly depleted by the action of these acids. In the presence of the enzymes, the fluorescence bleaching was dependent of chloride (Cl