Merck
  • Biomineralisation of fipronil and its major metabolite, fipronil sulfone, by Aspergillus glaucus strain AJAG1 with enzymes studies and bioformulation.
Biomineralisation of fipronil and its major metabolite, fipronil sulfone, by Aspergillus glaucus strain AJAG1 with enzymes studies and bioformulation.
3 Biotech (2017-07-02)
Anudurga Gajendiran, Jayanthi Abraham
ABSTRACT
Fipronil is a phenylpryazole insecticide which is extensively used for the protection of agricultural yields. However, this insecticide poses various threats to  the environment. Therefore it is essential to develop an effective method to degrade or eliminate this pollutant from the  environment. In this present study, a fungal strain AJAG1 capable of degrading fipronil and its metabolite, fipronil sulfone, was isolated through enrichment technique. Isolated fungal strain was identified as Aspergillus glaucus based upon its morphological, and 18S rRNA sequence analysis. Strain AJAG1 could degrade 900 mg L
Product Description
Supelco
Fipronil sulfone, PESTANAL®, analytical standard
Pricing and availability is not currently available.