Merck
  • Precocious centriole disengagement and centrosome fragmentation induced by mitotic delay.
Precocious centriole disengagement and centrosome fragmentation induced by mitotic delay.
Nature communications (2017-06-14)
Menuka Karki, Neda Keyhaninejad, Charles B Shuster
ABSTRACT
The spindle assembly checkpoint (SAC) delays mitotic progression until all sister chromatid pairs achieve bi-orientation, and while the SAC can maintain mitotic arrest for extended periods, moderate delays in mitotic progression have significant effects on the resulting daughter cells. Here we show that when retinal-pigmented epithelial (RPE1) cells experience mitotic delay, there is a time-dependent increase in centrosome fragmentation and centriole disengagement. While most cells with disengaged centrioles maintain spindle bipolarity, clustering of disengaged centrioles requires the kinesin-14, HSET. Centrosome fragmentation and precocious centriole disengagement depend on separase and anaphase-promoting complex/cyclosome (APC/C) activity, which also triggers the acquisition of distal appendage markers on daughter centrioles and the loss of procentriolar markers. Together, these results suggest that moderate delays in mitotic progression trigger the initiation of centriole licensing through centriole disengagement, at which point the ability to maintain spindle bipolarity becomes a function of HSET-mediated spindle pole clustering.
Sigma-Aldrich
Anti-PLK-4 Antibody, clone 6H5, clone 6H5, from mouse
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human ESPL1
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution