Merck
  • Restricting the induction of NGF in ovarian stroma engenders selective follicular activation through the mTOR signaling pathway.
Restricting the induction of NGF in ovarian stroma engenders selective follicular activation through the mTOR signaling pathway.
Cell death & disease (2017-05-26)
Yuanlin He, Xiaoxu Peng, Tinghe Wu, Weijie Yang, Wenwen Liu, Jing Zhang, Yiping Su, Feifei Kong, Xiaowei Dou, Jing Li
ABSTRACT
In mammalian ovaries, primordial follicles remain in a quiescent state until activation by the surrounding microenvironment. Ovarian intervention, for example, ovarian cystectomy, ovarian wedge resection or laser drilling therapies for polycystic ovarian syndrome, has long been reported to change follicular development by an unknown mechanism(s). Herein, we established a murine model with partial ovarian resection of one ovary unilaterally, with the contralateral ovary undamaged. We found the injury accelerated follicular activation and development through the mTORC1 signaling pathway. Moreover, the stimulation of primordial follicles was restricted near the incision site where the mTORC1 pathway showed sequential activation beginning at the interstitial cells and proceeding to the primordial follicles. Total and polysome-associated RNA-seq revealed the increase of the nerve growth factor (NGF) family member, in both two fractions and immunostaining showed the restricted induction of NGF near the incision site. In cultured newborn ovaries, NGF demonstrated increase of follicular activation, and moreover, the NGF inhibitor K252a effectively blocked activation of primordial follicles stimulated by the surgery. We liken ovulation in mammals to minor tissue trauma, which happens naturally and cyclically in the body. As the increase in NGF accompanied the accumulation of activated primordial follicles after ovulation, our study may represent a common mechanism for selective follicular activation induced by a localized increase in NGF in interstitial cells and mediated via the mTOR signaling pathway. In addition, the NGF inhibitor K252a and the mTOR inhibitor rapamycin constitute good candidates for protecting follicular reserve against over exhaustion after ovarian surgery.
Sigma-Aldrich
Ketamine hydrochloride/xylazine hydrochloride solution
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks