Merck
  • Leukocyte integrin Mac-1 regulates thrombosis via interaction with platelet GPIbα.
Leukocyte integrin Mac-1 regulates thrombosis via interaction with platelet GPIbα.
Nature communications (2017-05-31)
Yunmei Wang, Huiyun Gao, Can Shi, Paul W Erhardt, Alexander Pavlovsky, Dmitry A Soloviev, Kamila Bledzka, Valentin Ustinov, Liang Zhu, Jun Qin, Adam D Munday, Jose Lopez, Edward Plow, Daniel I Simon
ABSTRACT
Inflammation and thrombosis occur together in many diseases. The leukocyte integrin Mac-1 (also known as integrin α
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 392 mPa.s (25 °C)
Sigma-Aldrich
Glucose-6-phosphate Isomerase human, recombinant, expressed in E. coli
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)