Merck
  • Functional estimation of endothelin-1 receptor antagonism by bosentan, macitentan and ambrisentan in human pulmonary and radial arteries in vitro.
Functional estimation of endothelin-1 receptor antagonism by bosentan, macitentan and ambrisentan in human pulmonary and radial arteries in vitro.
European journal of pharmacology (2017-03-17)
James A Angus, Paul F Soeding, Richard J A Hughes, Christine E Wright
ABSTRACT
Endothelin receptor antagonists are approved for pulmonary arterial hypertension. Development of selective ET Human isolated pulmonary (i.d. 5.5mm) and human radial (i.d. 3.23mm) artery ring segments were mounted in organ baths for isometric force measurement. Single concentration-contraction curves to endothelin-1 were constructed in the absence or presence of bosentan (1-10µM), macitentan (0.03-0.3µM) or ambrisentan (0.1-1µM). All 3 endothelin antagonists caused competitive rightward shifts in the endothelin-1 concentration-response curves in both arteries. The Clark plot and analysis gave the following pK Noting the maximum plasma levels attained from recommended oral doses of each antagonist in volunteers, the pK
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
Ambrisentan, ≥98% (HPLC)