Merck
  • Development of europium doped core-shell silica cobalt ferrite functionalized nanoparticles for magnetic resonance imaging.
Development of europium doped core-shell silica cobalt ferrite functionalized nanoparticles for magnetic resonance imaging.
Acta biomaterialia (2016-12-06)
Bhavesh D Kevadiya, Aditya N Bade, Christopher Woldstad, Benson J Edagwa, JoEllyn M McMillan, Balasrinivasa R Sajja, Michael D Boska, Howard E Gendelman
ABSTRACT
The size, shape and chemical composition of europium (Eu A novel europium (Eu
Sigma-Aldrich
Europium(III) nitrate hydrate, 99.99% trace metals basis
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
1,2-Hexadecanediol, technical grade, 90%