- A versatile efficient one-step approach for carbazole-pyridine hybrid molecules: highly efficient host materials for blue phosphorescent OLEDs.
A versatile efficient one-step approach for carbazole-pyridine hybrid molecules: highly efficient host materials for blue phosphorescent OLEDs.
Chemical communications (Cambridge, England) (2014-12-17)
Chao Tang, Ran Bi, Youtian Tao, Fangfang Wang, Xudong Cao, Shifan Wang, Tao Jiang, Cheng Zhong, Hongmei Zhang, Wei Huang
PMID25500635
ABSTRACT
By using various fluoro-substituted pyridines and carbazole as starting materials, we have synthesized a series of carbazole-pyridine hybrid compounds through an efficient catalyst free C-N coupling reaction with high yields of 85-95%. The bi-, tri- and tetra-carbazole substituted pyridine derivatives exhibit increased thermal stabilities and comparatively high triplet energy levels of ∼3.0 eV. By using them as host materials for blue phosphorescent OLEDs, maximum current efficiencies are achieved ranging from 32-42 cd A(-1).