Merck
  • STIM1 Phosphorylation at Y361 Recruits Orai1 to STIM1 Puncta and Induces Ca
STIM1 Phosphorylation at Y361 Recruits Orai1 to STIM1 Puncta and Induces Ca
Scientific reports (2017-02-22)
Pascal Yazbeck, Mohammad Tauseef, Kevin Kruse, Md-Ruhul Amin, Rayees Sheikh, Stefan Feske, Yulia Komarova, Dolly Mehta
ABSTRACT
Store-operated Ca
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
MISSION® esiRNA, targeting human PTK2B
Sigma-Aldrich
O-Phospho-L-tyrosine