Merck
  • Liposomes for PET and MR Imaging and for Dual Targeting (Magnetic Field/Glucose Moiety): Synthesis, Properties, and in Vivo Studies.
Liposomes for PET and MR Imaging and for Dual Targeting (Magnetic Field/Glucose Moiety): Synthesis, Properties, and in Vivo Studies.
Molecular pharmaceutics (2016-12-29)
Jérémy Malinge, Bastien Géraudie, Paul Savel, Valérie Nataf, Aurélie Prignon, Claire Provost, Yongmin Zhang, Phalla Ou, Khaldoun Kerrou, Jean-Noël Talbot, Jean-Michel Siaugue, Matthieu Sollogoub, Christine Ménager
ABSTRACT
We describe the potentiality of a new liposomal formulation enabling positron emission tomography (PET) and magnetic resonance MR() imaging. The bimodality is achieved by coupling a
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)