Merck
  • Novel Bioactive Co(II), Cu(II), Ni(II) and Zn(II) Complexes with Schiff Base Ligand Derived from Histidine and 1,3-Indandione: Synthesis, Structural Elucidation, Biological Investigation and Docking Analysis.
Novel Bioactive Co(II), Cu(II), Ni(II) and Zn(II) Complexes with Schiff Base Ligand Derived from Histidine and 1,3-Indandione: Synthesis, Structural Elucidation, Biological Investigation and Docking Analysis.
Journal of fluorescence (2016-10-04)
V Violet Dhayabaran, T Daniel Prakash, R Renganathan, Elsa Friehs, Detlef W Bahnemann
ABSTRACT
Novel bioactive complexes of Co(II), Cu(II), Ni(II) and Zn(II) metal ions with Schiff base ligand derived from histidine and 1,3-indandione were synthesized and thoroughly characterized by various analytical and spectral techniques. The biological investigations were carried out to examine the efficiency of the binding interaction of all the complexes with calf thymus DNA (CT-DNA). The binding properties were studied and evaluated quantitatively by K
Sigma-Aldrich
1,3-Indandione, 97%