Merck
  • A comparative study on structural elucidation of sialyl oligosaccharides by mass spectrometry with fast atom bombardment, electrospray ionization, and matrix assisted laser desorption/ionization.
A comparative study on structural elucidation of sialyl oligosaccharides by mass spectrometry with fast atom bombardment, electrospray ionization, and matrix assisted laser desorption/ionization.
Bioscience, biotechnology, and biochemistry (2002-01-17)
M Okamoto
ABSTRACT
To establish a new protocol for sensitive detection and structural characterization of sialyl oligosaccharides, their sensitivities and structural information from mass spectrometry and tandem mass spectrometry with FAB-, ESI-, and MALDI were evaluated in detail. Among these ionization methods, FAB-MS and FAB-MS/MS gave reproducible and predictable spectra carrying information on sequence and branching of sialyl oligosaccharides after derivatization with 2-aminopyridine (PA). With both positive and negative ion modes, their structural elucidation promises to be straightforward, MS/MS spectra being measurable at as low as 200 pmol. Thus, this method constitutes a powerful tool for sensitive detection and structural characterization of limited quantities of sialyl oligosaccharides by FAB-MS and FAB-MS/MS.
Sigma-Aldrich
2-Aminopyridine, purum, ≥98.0% (NT)