Merck
  • Influence of storage and heating on protein glycation levels of processed lactose-free and regular bovine milk products.
Influence of storage and heating on protein glycation levels of processed lactose-free and regular bovine milk products.
Food chemistry (2016-12-17)
Sanja Milkovska-Stamenova, Ralf Hoffmann
ABSTRACT
Thermal treatment preserves the microbiological safety of milk, but also induces Maillard reactions modifying for example proteins. The purpose of this study was evaluating the influence of consumer behaviors (storage and heating) on protein glycation degrees in bovine milk products. Lactosylation and hexosylation sites were identified in ultra-high temperature (UHT), lactose-free pasteurized, and lactose-free UHT milk (ULF) and infant formula (IF) using tandem mass spectrometry (electron transfer dissociation). Overall, 303 lactosylated and 199 hexosylated peptides were identified corresponding to 170 lactosylation (31 proteins) and 117 hexosylation sites (25 proteins). In quantitative terms, storage increased lactosylation up to fourfold in UHT and IF and hexosylation up to elevenfold in ULF and threefold in IF. These levels increased additionally twofold when the stored samples were heated (40°C). In conclusion, storage and heating appear to influence protein glycation levels in milk at similar or even higher degrees than industrial processing.
Sigma-Aldrich
CHAPS 100 mM solution
Pricing and availability is not currently available.