Merck
  • A54 Peptide Modified and Redox-Responsive Glucolipid Conjugate Micelles for Intracellular Delivery of Doxorubicin in Hepatocarcinoma Therapy.
A54 Peptide Modified and Redox-Responsive Glucolipid Conjugate Micelles for Intracellular Delivery of Doxorubicin in Hepatocarcinoma Therapy.
ACS applied materials & interfaces (2016-12-10)
Na Liu, Yanan Tan, Yingwen Hu, Tingting Meng, Lijuan Wen, Jingwen Liu, Bolin Cheng, Hong Yuan, Xuan Huang, Fuqiang Hu
ABSTRACT
Redox-responsive nanomaterials applied in drug delivery systems (DDS) have attracted an increasing attention in pharmaceutical research as a carrier for antitumor therapy. However, there would be unwanted drug release from a redox-responsive DDS with no selection at nontarget sites, leading to undesirable toxicities in normal tissues and cells. Here, an A54 peptide modified and PEGylated reduction cleavable glucolipid conjugate (A54-PEG-CSO-ss-SA, abbreviated to APCssA) was designed for intracellular delivery of doxorubicin (DOX). The synthesized APCssA could be assembled via micellization self-assembly in aqueous water above the critical micelle concentration (54.9 μg/mL) and exhibited a high drug encapsulation efficiency (77.92%). The APCssA micelles showed an enhanced redox sensitivity in that the disulfide bond could be degraded quickly and the drug would be released from micelles in 10 mM levels of glutathione (GSH). The cellular uptake studies highlighted the affinity of APCssA micelles toward the hepatoma cells (BEL-7402) compared to that toward HepG2 cells. In contrast with the nonresponsive conjugate, the drug was released from APCssA micelles more quickly in 10 mM level of GSH concentration (tumor cells). Moreover, the DOX-loaded APCssA micelles displayed an increased cytotoxicity which was 1.6- to 2.0-fold that of unmodified and nonresponsive micelles. In vivo, the APCssA micelles had stronger distribution to liver and hepatoma tissue and prolonged the circulation and retention time, while the drug release only occurred in the tumor tissue. The APCssA/DOX showed the tumor inhibition rate equal to that of commercial doxorubicin hydrochloric without negative consequence. This study suggested that the APCssA/DOX showed promising potential to treat the tumor for its special tumor targeting, selective intracellular drug release, enhanced antitumor activity, and reduced toxicity on normal tissues.
Sigma-Aldrich
3,3′-Dithiodipropionic acid, 95%